Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hsp70 interacting protein HIP Rabbit anti-Human, Mouse, Clone: 4A3G8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Hsp70 interacting protein HIP |
---|---|
Clone | 4A3G8 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Hsp70 interacting protein HIP Monoclonal antibody specifically detects Hsp70 interacting protein HIP in Human, Mouse samples. It is validated for Western BlotSpecifications
Hsp70 interacting protein HIP | |
Western Blot | |
Unconjugated | |
Rabbit | |
Lipid and Metabolism, Membrane Trafficking and Chaperones | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6767 | |
IgG | |
Affinity purified |
4A3G8 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein | |
A synthetic peptide corresponding to a sequence within amino acids 270-369 of human Hsp70 interacting protein HIP (P50502). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title