Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hsp70 interacting protein HIP Rabbit anti-Human, Mouse, Rat, Bovine, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.00 - $611.00
Specifications
Antigen | Hsp70 interacting protein HIP |
---|---|
Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB12013490
![]() |
Novus Biologicals
NB12013490 |
50 μg |
Each for $342.00
|
|
|||||
NB120134902
![]() |
Novus Biologicals
NB12013490200UG |
200 μg |
Each for $611.00
|
|
|||||
NB018068
![]() |
Novus Biologicals
NB120134900.025MG |
0.025 mg |
Each for $182.00
|
|
|||||
Description
Hsp70 interacting protein HIP Polyclonal specifically detects Hsp70 interacting protein HIP in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Hsp70 interacting protein HIP | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Membrane Trafficking and Chaperones | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
6767 | |
Recombinant Histidine 6 fusion protein (Rat). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Bovine | |
AAG2, aging-associated protein 2, FAM10A1FAM10A4, heat shock 70kD protein binding protein, Hip, HIPFLJ27260, HOP, hsc70-interacting protein, Hsp70-interacting protein, HSPABP1, P48MGC129952, PRO0786, Progesterone receptor-associated p48 protein, Protein FAM10A1, Putative tumor suppressor ST13, Renal carcinoma antigen NY-REN-33, SNC6HSPABP, suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein), suppression of tumorigenicity 13 (colon carcinoma) (Hsp70-interacting protein), Suppression of tumorigenicity 13 protein | |
ST13 | |
IgG | |
Protein A purified | |
This detects both recombinant rat HSC70 Interacting Protein HIP as well as the native protein in tissue extracts. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title