Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp70 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277426
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7426 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7426 Supplier Leinco Technologies Inc Supplier No. 11147100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Hsp70 Polyclonal antibody specifically detects Hsp70 in Invertebrate samples. It is validated for Western Blot

Specifications

Antigen Hsp70
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Alias 68 kDa heat shock protein; 70 kda heat shock protein-2; 70 kilodalton heat shock protein; 87A; 87A locus; 87C locus; Bb; CG18743; CG31359; CG31359-PA; CG31366; CG31366-PA; CG31449; CG6489; chaperone; dHSP70; dhsp70Aa; Dmel\CG31359; Dmel\CG31366; Dmel_CG31359; Dmel_CG31366; Dm-hsp70; DmHSP70AA; DMHSP7D1; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; Hcp70.1; heat shock 10kDa protein 1-like; heat shock 70; heat shock 70 kD protein 1; heat shock 70 kD protein 2; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 1-Hom; Heat shock 70 kDa protein 1L; heat shock 70 kDa protein 1-like; heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70 kDa-like protein 1; heat shock 70kD protein 1A; heat shock 70kD protein 1B; heat shock 70kD protein 1B (mapped); heat shock 70kD protein-like 1; heat shock 70kDa protein 1A; heat shock 70kDa protein 1B; heat shock 70kDa protein 1-like; heat shock cognate 70-kd protein, tandem duplicate 2; Heat shock protein; heat shock protein 1A; heat shock protein 1B; heat shock protein 1-like; heat shock protein 70; heat shock protein 70.1; heat shock protein 70.2; heat shock protein 70-1; Heat shock protein 70-1l; Heat shock protein 70Aa; Heat shock protein 70Ab; Heat shock protein 70Ba; Heat shock protein 70Bb; Heat shock protein 70Bc; heat shock protein cognate 70, testis; heat shock protein family A (Hsp70) member 1 like; heat shock protein family A (Hsp70) member 1A; heat shock protein family A (Hsp70) member 1B; heat shock protein, 70 kDa 1; heat shock protein, 70 kDa 3; heat shock protein-70; heat shock-induced protein; heat shock-related protein hsc70t; heat-shock 70-kilodalton protein 1A; heat-shock 70-kilodalton protein 1B; heat-shock protein 70; Heat-shock-protein-70Aa; Heat-shock-protein-70Bb; HEL S 103; HEL-S-103; hs70; Hsc70t; HSP; Hsp 70; hsp68; Hsp70; Hsp-70; HSP70 1; HSP70 1B; HSP70 2; hsp70 87A; hsp70 87C; hsp70 Aa; HSP70 AceyltK77; hsp70(87A); Hsp70(87C); HSP70.1; HSP70.1/2; HSP70.1/HSP70.2; HSP70.2; Hsp70.3; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; HSP70-1B; HSP70-1L; HSP70-2; Hsp70-3; HSP70-87A7; HSP70-87C1; hsp70a; hsp70A1; Hsp70A7d; Hsp70Aa; Hsp-70Aa; Hsp70Aa-PA; Hsp70Ab; Hsp70B; Hsp70Ba; Hsp70Bb; Hsp70Bbc; Hsp70Bb-PA; Hsp70Bc; HSP70-HOM; HSP70I; HSP70T; HSP72; Hspa1; Hspa1a; HSPA1B; Hspa1l; Hspa2; HSPA4; Hspa70; HSX70; hum70t; inducible heat shock protein 70; LP05203p; LP05233p; LP08776p; Major heat shock 70 kDa protein Aa; Major heat shock 70 kDa protein Ab; Major heat shock 70 kDa protein Ba; Major heat shock 70 kDa protein Bb; Major heat shock 70 kDa protein Bc; Msh5; protein 70; si:ch73-46h18.6; si:dkeyp-69e1.1; Spermatid-specific heat shock protein 70; zgc:174006
Host Species Rabbit
Immunogen His-tagged C-terminal amino acids 365-681 of P. falciparum Hsp70 (Accession no. M19753).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Invertebrate
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.