Learn More
Invitrogen™ HSP70 Recombinant Rabbit Monoclonal Antibody (SA0379)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA531961
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
HSP70 is a Heat shock protein (HSP) which are expressed in response to various biological stresses, including high temperatures. There are several major families of HSPs that include HSP70, there are HSP90 and HSP100. The HSP70 family is a set of highly conserved proteins that are induced by a variety of biological stresses, including heat stress, in every organism in which the proteins have been examined. The human HSP70 family members include: HSP70, a protein which is strongly inducible in all organisms but which is also constitutively expressed in primate cells; HSP72, a 72 kDa protein that is induced exclusively under stress conditions; HSC70, or cognate protein, is a 72 kDa, constitutively expressed, protein which is involved in the uncoating of clathrin coated vesicles; GRP78, or BiP, is a glucose regulated 78 kDa protein localized in the endoplasmic reticulum; and p75, or HSP75, a 75 kDa protein that is found within the mitochondria. Further, HSP70 is encoded by an intronless gene and, in conjunction with other heat shock proteins, HSP70 stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and organelles. HSP70 is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The HSP70 gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
Specifications
HSP70 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
O88600, P0DMV8, P0DMV9, P0DMW0, P0DMW1, P14659, P16627, P17156, P17879, P34931, P34932, P54652, P55063, Q61316, Q61696 | |
HSPA1A, HSPA1B, Hspa1l, HSPA2, HSPA4 | |
Recombinant protein within Human Hsp70 aa 403-641. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
SA0379 | |
Unconjugated | |
HSPA1B | |
68 kDa heat shock protein; 70 kda heat shock protein-2; 70 kilodalton heat shock protein; 87A; 87A locus; 87C locus; Bb; CG18743; CG31359; CG31359-PA; CG31366; CG31366-PA; CG31449; CG6489; chaperone; dHSP70; dhsp70Aa; Dmel\CG31359; Dmel\CG31366; Dmel_CG31359; Dmel_CG31366; Dm-hsp70; DmHSP70AA; DMHSP7D1; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; Hcp70.1; heat shock 10kDa protein 1-like; heat shock 70; heat shock 70 kD protein 1; heat shock 70 kD protein 2; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 1-Hom; Heat shock 70 kDa protein 1L; heat shock 70 kDa protein 1-like; heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70 kDa-like protein 1; heat shock 70kD protein 1A; heat shock 70kD protein 1B; heat shock 70kD protein 1B (mapped); heat shock 70kD protein-like 1; heat shock 70kDa protein 1A; heat shock 70kDa protein 1B; heat shock 70kDa protein 1-like; heat shock cognate 70-kd protein, tandem duplicate 2; Heat shock protein; heat shock protein 1A; heat shock protein 1B; heat shock protein 1-like; heat shock protein 70; heat shock protein 70.1; heat shock protein 70.2; heat shock protein 70-1; Heat shock protein 70-1l; Heat shock protein 70Aa; Heat shock protein 70Ab; Heat shock protein 70Ba; Heat shock protein 70Bb; Heat shock protein 70Bc; heat shock protein cognate 70, testis; heat shock protein family A (Hsp70) member 1 like; heat shock protein family A (Hsp70) member 1A; heat shock protein family A (Hsp70) member 1B; heat shock protein, 70 kDa 1; heat shock protein, 70 kDa 3; heat shock protein-70; heat shock-induced protein; heat shock-related protein hsc70t; heat-shock 70-kilodalton protein 1A; heat-shock 70-kilodalton protein 1B; heat-shock protein 70; Heat-shock-protein-70Aa; Heat-shock-protein-70Bb; HEL S 103; HEL-S-103; hs70; Hsc70t; HSP; Hsp 70; hsp68; Hsp70; Hsp-70; HSP70 1; HSP70 1B; HSP70 2; hsp70 87A; hsp70 87C; hsp70 Aa; HSP70 AceyltK77; hsp70(87A); Hsp70(87C); HSP70.1; HSP70.1/2; HSP70.1/HSP70.2; HSP70.2; Hsp70.3; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; HSP70-1B; HSP70-1L; HSP70-2; Hsp70-3; HSP70-87A7; HSP70-87C1; hsp70a; hsp70A1; Hsp70A7d; Hsp70Aa; Hsp-70Aa; Hsp70Aa-PA; Hsp70Ab; Hsp70B; Hsp70Ba; Hsp70Bb; Hsp70Bbc; Hsp70Bb-PA; Hsp70Bc; HSP70-HOM; HSP70I; HSP70T; HSP72; Hspa1; Hspa1a; HSPA1B; Hspa1l; Hspa2; HSPA4; Hspa70; HSX70; hum70t; inducible heat shock protein 70; LP05203p; LP05233p; LP08776p; Major heat shock 70 kDa protein Aa; Major heat shock 70 kDa protein Ab; Major heat shock 70 kDa protein Ba; Major heat shock 70 kDa protein Bb; Major heat shock 70 kDa protein Bc; Msh5; protein 70; si:ch73-46h18.6; si:dkeyp-69e1.1; Spermatid-specific heat shock protein 70; zgc:174006 | |
Rabbit | |
Protein A | |
RUO | |
15482, 15511, 15512, 15525, 193740, 24472, 24963, 266759, 294254, 3303, 3304, 3305, 3306, 3308, 60460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.