Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP701L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595961
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595961 Supplier Invitrogen™ Supplier No. PA595961
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STRIPKVQRL LQDYFNGRDL NKSINPDEAV AYGAAVQAAI LMGDKSEKVQ DLLLLDVAPL SLGLETAGGV MTALIKRNST IPTKQTQIFT TYSDNQPGVL IQVYEGERAM TKDNNLLGRF DLTGIPPAPR GVPQIEVTFD IDANGILNVT ATDKSTGKVN KITITNDKGR LSKEEIERMV LDAEKYKAED EVQREKIAAK NALESYAFNM KSVVSDEGLK GKISESDKNK ILDKCNELLS WLEVNQLAEK DEFDHKRKEL EQMCNPIITK LYQGGCTGPA CGTGYVPGRP ATGPTIEEVD; Positive Samples: HeLa, THP-1, ES-2, Mouse testis.

In response to adverse changes in their environment, cells from many organisms increase the expression of a class of proteins referred to as heat shock proteins. One class of stress proteins, termed the small heat shock proteins is comprised of a diverse group of proteins from approximately15 to >30 kDa, with a single form found in yeast and human cells and multiple related forms found in higher plants. A direct relationship between elevated strwess protein expression and neoplasia has been suggested from studies on estrogen-induced proteins in breast cancer cells. In addition, Hsp27 exhibits rapid increased phosphorylation in response to various mitogens, tumor promoter (e. g. phorbol esters) and calcium ionophores. The expression of a 24 kDa protein was shown to correlate with steroid hormone receptor in cell lines, organs, and tumors. The 24 kDa estrogen-induced protein was found to be expressed at high levels in carcinoma of the breast. Another study has suggested that levels of the Hsp27 correlate with endometrial adenocarcinomas. Interestingly, the regulation of human Hsp27 by estrogens and heat shock is reminiscent of the ecdysteroid and heat shock activation of the drosophila Hsp20 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP701L
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Hspa1l
Gene Accession No. P16627, P34931
Gene Alias heat shock 10kDa protein 1-like; heat shock 70 kDa protein 1-Hom; heat shock 70 kDa protein 1L; Heat shock 70 kDa protein 1-like; Heat shock 70 kDa protein 3; heat shock 70 kDa-like protein 1; heat shock 70kD protein-like 1; heat shock 70kDa protein 1-like; Heat shock protein; heat shock protein 1-like; Heat shock protein 70-1l; heat shock protein cognate 70, testis; heat shock protein family A (Hsp70) member 1 like; heat shock-related protein hsc70t; Hsc70t; HSP70.3; HSP70-1L; Hsp70-3; HSP70-HOM; HSP70T; Hspa1l; hum70t; inducible heat shock protein 70; Msh5; Spermatid-specific heat shock protein 70
Gene Symbols Hspa1l
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 342-641 of human HSPA1L (NP_0055183).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15482, 3305
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.