Learn More
Invitrogen™ HSP90 alpha Monoclonal Antibody (2G5.G3), FITC
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545181
Description
0.5 μg/mL of MA5-45181 was sufficient for detection of HSP90alpha in 20 μg of heat shocked HeLa cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 90kDa. HSP90 alpha -specific (>96% alpha -specific by ELISA).
Heat shock proteins (HSP) are proteins that are expressed in response to various biological stress, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, and some kinases. It has been suggested that HSP90 is necessary to maintain structural integrity of at least some steroid receptors to allow for ligand binding during receptor activation.
Specifications
HSP90 alpha | |
Monoclonal | |
1 mg/mL | |
PBS, 9.1mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 | |
P07900, P07901, P82995 | |
Hsp90aa1 | |
Recombinant Full Length Human HSP90alpha Protein. | |
200 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
2G5.G3 | |
FITC | |
Hsp90aa1 | |
86kDa; 89kDa; 90-kDa heat shock protein; 90-kDa heat shock protein alpha; akineto; AL024080; AL024147; EL52; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; fb17b01; heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 86; heat shock protein 90; heat shock protein 90 alpha family class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1, tandem duplicate 1; heat shock protein 90-alpha 1; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein alpha; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-alpha 1; heat shock protein HSP90 alpha; heat shock protein hsp90alpha; heat shock protein, 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; Heat shock protein90; HEL-S-65p; HSP; HSP 86; HSP 90 alpha; hsp4; HSP86; Hsp86-1; Hsp89; HSP89A; HSP90; hsp90a; hsp90a protein; hsp90a.1; hsp90aa1; hsp90aa1.1; hsp90alpha; HSP90N; HSPC1; Hspca; HSPCA protein; HSPCAL1; HSPCAL4; HSPN; hypothetical protein; LAP2; LAP-2; Lipopolysaccharide-associated protein 2; LPS-associated protein 2; renal carcinoma antigen NY-REN-38; slo; sloth; TSTA; Tumor-specific transplantation 86 kDa antigen; wu:fb17b01; zgc:86652 | |
Mouse | |
Protein G | |
RUO | |
15519, 299331, 3320 | |
4°C, store in dark | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.