Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp90 alpha Mouse anti-Human, Unconjugated, Clone: 1G6-D7, MilliporeSigma™

Catalog No. MABC110625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1106-25 25 μg
MABC1106-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1106-25 Supplier MilliporeSigma Supplier No. MABC110625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Hsp90 alpha, clone 1G6-D7, Cat. No. MABC1106, is a mouse monoclonal antibody that detects Hsp90 alpha, Hsp90 beta and has been tested for use in ELISA, Immunoprecipitation, Functional assay, Inhibition, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp90 alpha
Applications ELISA, Immunoprecipitation, Functional Assay, Inhibition Assays, Western Blot
Classification Monoclonal
Clone 1G6-D7
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias Heat shock protein HSP 90-alpha;Heat shock 86 kDa;HSP 86;Lipopolysaccharide-associated protein 2;LAP-2;LPS-associated protein 2;Renal carcinoma antigen NY-REN-38
Gene Symbols HSP90AA1;HSP90A;HSPC1;HSPCA
Host Species Mouse
Immunogen A recombinant fragment corresponding to 115 amino acids from the F-5 region of human HSP90 alpha.
Molecular Weight of Antigen ∽90 kDa observed; 84.66 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001017963.2
Test Specificity Clone 1G6-D7 is a mouse monoclonal antibody that detects HSP90 alpha and HSP90 beta in human cells. it targets an epitope within 9 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.