Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP90 alpha Rabbit anti-Human, Mouse, Rat, Clone: 0R3S6, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31652820UL
This item is not returnable.
View return policy
Description
HSP90 alpha Monoclonal antibody specifically detects HSP90 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| HSP90 alpha | |
| Monoclonal | |
| Unconjugated | |
| FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HSP90 alpha (P07900). | |
| 20 μg | |
| Cancer, Chaperone Mediated Autophagy (CMA), Hypoxia, Membrane Trafficking and Chaperones | |
| 3320 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0R3S6 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction