Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP90 alpha Recombinant Rabbit Monoclonal Antibody (4B5)
GREENER_CHOICE

Catalog No. PIMA533173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533173 Supplier Invitrogen™ Supplier No. MA533173
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

HSP90 alpha Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA

Heat shock proteins (HSP) are proteins that are expressed in response to various biological stress, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, and some kinases. It has been suggested that HSP90 is necessary to maintain structural integrity of at least some steroid receptors to allow for ligand binding during receptor activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP90 alpha
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot
Classification Recombinant Monoclonal
Clone 4B5
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Hsp90aa1
Gene Accession No. P07900, P82995
Gene Alias 86kDa; 89kDa; 90-kDa heat shock protein; 90-kDa heat shock protein alpha; akineto; AL024080; AL024147; EL52; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; fb17b01; heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 86; heat shock protein 90; heat shock protein 90 alpha family class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1; heat shock protein 90, alpha (cytosolic), class A member 1, tandem duplicate 1; heat shock protein 90-alpha 1; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein alpha; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-alpha 1; heat shock protein HSP90 alpha; heat shock protein hsp90alpha; heat shock protein, 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; Heat shock protein90; HEL-S-65p; HSP; HSP 86; HSP 90 alpha; hsp4; HSP86; Hsp86-1; Hsp89; HSP89A; HSP90; hsp90a; hsp90a protein; hsp90a.1; hsp90aa1; hsp90aa1.1; hsp90alpha; HSP90N; HSPC1; Hspca; HSPCA protein; HSPCAL1; HSPCAL4; HSPN; hypothetical protein; LAP2; LAP-2; Lipopolysaccharide-associated protein 2; LPS-associated protein 2; renal carcinoma antigen NY-REN-38; slo; sloth; TSTA; Tumor-specific transplantation 86 kDa antigen; wu:fb17b01; zgc:86652
Gene Symbols Hsp90aa1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSP90AA1 (aa 2-12).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299331, 3320
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.