Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90 Antibody (AC88), Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 6 publications

$206.50 - $529.95

Specifications

Antigen HSP90
Clone AC88
Concentration 1.0 mg/mL
Dilution Western Blot 1 ug/ml, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017429
SDP
View Documents
Novus Biologicals
NB10019720.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB1001972
SDP
View Documents
Novus Biologicals
NB1001972
0.1 mg
Each for $529.95
Only null left
Add to Cart
 
Description

Description

HSP90 Monoclonal specifically detects HSP90 in Human, Mouse, Rat, Porcine, Bacteria, Canine, Chicken, Fish, Guinea Pig, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular).
Specifications

Specifications

HSP90
1.0 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry, Flow Cytometry
Unconjugated
Mouse
Rabbit, Sheep, Rat, Primate, Mouse, Bacteria, Fish, Chicken, Human, Canine, Pig, Guinea Pig
3320
Achlya ambisexualis (water mold) Hsp90.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
90 kDa
AC88
Western Blot 1 ug/ml, Flow Cytometry, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Flow (Intracellular)
Monoclonal
Purified
RUO
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38
HSP90AA1
IgG1
Protein G purified
Recognizes both Hsp90 alpha and beta. It reacts with the rodent glucocorticoid receptor and with proliferation potential proteins (P2PS) with apparent molecular masses of 30-40 kDa.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.