Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP90 Antibody (D7A), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11096872FR
Description
HSP90 Monoclonal specifically detects HSP90 in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HSP90 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
Mouse | |
Protein A or G purified | |
Cancer, Hypoxia, Membrane Trafficking and Chaperones | |
3320 | |
Chicken | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
D7A | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
P11501 | |
HSP90AA1 | |
Full length protein HSP90 purified from chicken brain | |
0.1 mL | |
Primary | |
Recognizes 90kDa. Can isolate complexes of HSP90, Src kinase and cec37. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction