Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSP90 beta Monoclonal Antibody (MBH90B)
GREENER_CHOICE

Catalog No. PIMA110373
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA110373 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA110373 Supplier Invitrogen™ Supplier No. MA110373
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes the EEVHHG epitope within the N-terminal part of Hsp90 beta an ubiquitously expressed intracellular protein with calculated Mw of 83.3 kDa, however, migrating as a 90 kDa band under reducing SDS-PAGE conditions.

Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSP90 beta
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone MBH90B
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene Hsp90ab1
Gene Accession No. P08238, P11499, Q76LV1
Gene Alias 90kDa; 90-kDa heat shock protein beta; AL022974; C81438; D6S182; heat shock 84 kDa; heat shock 90 kDa protein; heat shock 90kD protein 1, beta; heat shock 90kDa protein 1, beta; heat shock cognate protein HSP 90-beta; Heat shock protein; heat shock protein 1, beta; heat shock protein 90 alpha (cytosolic), class B member 1; heat shock protein 90 alpha family class B member 1; heat shock protein 90 beta; heat shock protein 90 kDa; heat shock protein 90 kDa alpha class B member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-beta; heat shock protein, 84 kDa 1; HSP; HSP 84; HSP 90; Hsp84; Hsp84-1; Hsp90; hsp90 beta; HSP90AB1; HSP90B; HSP90-beta; HSPC2; HSPCB; retinal degeneration slow protein; RP1-302G2.1; TSTA; Tumor-specific transplantation 84 kDa antigen; Unknown (protein for IMAGE:7943063)
Gene Symbols Hsp90ab1
Host Species Mouse
Immunogen Peptide corresponding to the EEVHHGEEEVEC sequence within N-terminal part of human Hsp90 (aa 3-14).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15516, 3326, 767874
Target Species Human, Mouse, Bovine
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.