Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp90 Monoclonal Antibody (4F3.E8), QED Bioscience™
SDP

Catalog No. 502277404
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7404 Supplier Leinco Technologies Inc Supplier No. 11113100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hsp90 Monoclonal antibody specifically detects Hsp90 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot

Specifications

Antigen Hsp90
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4F3.E8
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene HSP90AA1
Gene Accession No. P07900, P07901, P08238, P11499, P34058, P82995
Gene Alias 86kDa; 89kDa; 90kDa; 90-kDa heat shock protein; 90-kDa heat shock protein alpha; 90-kDa heat shock protein beta; AL022974; AL024080; AL024147; C81438; D6S182; EL52; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; heat shock 84 kDa; Heat shock 86 kDa; heat shock 90 kDa protein; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein 1, beta; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 1, beta; heat shock protein 86; heat shock protein 90 alpha (cytosolic), class B member 1; heat shock protein 90 alpha family class A member 1; heat shock protein 90 alpha family class B member 1; heat shock protein 90 beta; heat shock protein 90 kDa; heat shock protein 90, alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein 90kDa alpha family class B member 1; heat shock protein HSP 90-alpha; Heat shock protein HSP 90-beta; heat shock protein, 1; heat shock protein, 84 kDa 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; HEL-S-65p; HSP; HSP 84; HSP 86; HSP 90; hsp4; HSP84; Hsp84-1; HSP86; Hsp86-1; Hsp89; HSP89A; Hsp90; HSP90A; Hsp90aa1; Hsp90ab1; HSP90B; HSP90-BETA; HSP90N; HSPC1; HSPC2; HSPCA; HSPCA protein; HSPCAL1; HSPCAL4; HSPCB; HSPN; LAP2; LAP-2; lipopolysaccharide-associated protein 2; LPS-associated protein 2; renal carcinoma antigen NY-REN-38; retinal degeneration slow protein; TSTA; Tumor-specific transplantation 84 kDa antigen; tumor-specific transplantation 86 kDa antigen; Unknown (protein for IMAGE:7943063)
Host Species Mouse
Immunogen Recombinant human Hsp90 purified from E. coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15516, 15519, 299331, 301252, 3320, 3326
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.