Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp90 Mouse anti-Human, Clone: 3C9, Bioss
SDP

Catalog No. 501979832
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9832 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9832 Supplier Bioss Supplier No. BSM51215M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. There are 2 major cytosolic HSP90 proteins, HSP90AA1, an inducible form, and HSP90AB1 and mitochondria (Chen et al., 2005 ).

Specifications

Antigen Hsp90
Applications Western Blot
Classification Monoclonal
Clone 3C9
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene HSP90AB1
Gene Accession No. P07900, P08238
Gene Alias 86kDa; 89kDa; 90kDa; 90-kDa heat shock protein; AL022974; AL024080; AL024147; BOS_20461; BOS_21524; C81438; D6S182; EL52; epididymis luminal secretory protein 52; epididymis secretory sperm binding protein Li 65p; Heat shock 84 kDa; Heat shock 86 kDa; heat shock 90kD protein 1, alpha; heat shock 90kD protein 1, alpha-like 4; heat shock 90kD protein 1, beta; heat shock 90kD protein, alpha-like 4; heat shock 90kDa protein 1, alpha; heat shock 90kDa protein 1, beta; Heat shock protein; heat shock protein 1, alpha; heat shock protein 1, beta; heat shock protein 86; heat shock protein 90 alpha (cytosolic), class B member 1; heat shock protein 90 alpha family class A member 1; heat shock protein 90 alpha family class B member 1; heat shock protein 90 kDa; heat shock protein 90, alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha (cytosolic), class A member 1; heat shock protein 90kDa alpha (cytosolic), class B member 1; heat shock protein 90kDa alpha family class A member 1; heat shock protein 90kDa alpha family class B member 1; Heat shock protein HSP 90-alpha; Heat shock protein HSP 90-beta; heat shock protein, 1; heat shock protein, 84 kDa 1; heat shock protein, 86 kDa 1; heat shock protein, 89 kDa; HEL-S-65p; HSP; HSP 84; HSP 86; HSP 90; hsp4; Hsp84; Hsp84-1; Hsp86; Hsp86-1; Hsp89; HSP89A; HSP90; HSP90A; HSP90AA1; Hsp90ab1; HSP90B; HSP90-BETA; HSP90N; HSPC1; HSPC2; HSPCA; HSPCAL1; HSPCAL4; HSPCB; HSPN; LAP2; LAP-2; Lipopolysaccharide-associated protein 2; LOC100338694; LOC100358690; LPS-associated protein 2; Renal carcinoma antigen NY-REN-38; retinal degeneration slow protein; TSTA; Tumor-specific transplantation 84 kDa antigen; Tumor-specific transplantation 86 kDa antigen
Gene Symbols HSP90AA1, HSP90AB1
Host Species Mouse
Immunogen This antibody is generated from a mouse immunized with a KLH conjugated synthetic peptide between 150-400 amino acids from human.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3320, 3326
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.