Learn More
Description
The 90kDa heat shock protein (Hsp90), a highly conserved stress-induced protein, is abundantly expressed in most tissues under nonstress conditions and is required for eukaryotic cell viability. Hsp90 interacts with a number of signaling molecules, other heat shock proteins, and cytoskeletal proteins. However, the function of Hsp90 in these complexes remains unclear. It is thought that Hsp90 plays a common role in a number of diverse signaling systems due to its physical association with v-Src family tyrosine kinases, steroid hormone receptors, the Raf serine/threonine kinase, the basic-helix-loop-helix dioxin receptor, and the βγ subunits of G proteins.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Specifications
| Antigen | Hsp90 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 68 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Description | Heat Shock Protein-90 |
| Formulation | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.