Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsp90 Mouse, Unlabeled, Clone: 68, BD

Catalog No. BDB610419 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610419 150 μg
BDB610418 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610419 Supplier BD Biosciences Supplier No. 610419
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 90kDa heat shock protein (Hsp90), a highly conserved stress-induced protein, is abundantly expressed in most tissues under nonstress conditions and is required for eukaryotic cell viability. Hsp90 interacts with a number of signaling molecules, other heat shock proteins, and cytoskeletal proteins. However, the function of Hsp90 in these complexes remains unclear. It is thought that Hsp90 plays a common role in a number of diverse signaling systems due to its physical association with v-Src family tyrosine kinases, steroid hormone receptors, the Raf serine/threonine kinase, the basic-helix-loop-helix dioxin receptor, and the βγ subunits of G proteins.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hsp90
Applications Western Blot
Classification Monoclonal
Clone 68
Concentration 250μg/mL
Conjugate Unconjugated
Description Heat Shock Protein-90
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.