Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hsp90A/B Rabbit anti-Human, Mouse, Rat, Clone: 5D7Y6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Hsp90A/B |
---|---|
Clone | 5D7Y6 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Hsp90A/B Monoclonal antibody specifically detects Hsp90A/B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Hsp90A/B | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Endocrinology, Immunology, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3320 | |
IgG | |
Affinity purified |
5D7Y6 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
FLJ31884, Heat shock 86 kDa, heat shock 90kD protein 1, alpha, heat shock 90kD protein 1, alpha-like 4, heat shock 90kD protein, alpha-like 4, heat shock 90kDa protein 1, alpha, heat shock protein 90kDa alpha (cytosolic), class A member 1, heat shock protein HSP 90-alpha, Hsp89, Hsp90, HSP90AHSP86, HSP90N, HSPC1HSP 86, HSPCAHSP89A, HSPCAL1, HSPCAL4, HSPN, LAP2, Renal carcinoma antigen NY-REN-38 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 533-732 of human Hsp90A/B (P07900). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title