Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90AA1/HSP90AB1 Mouse anti-Human, Biotin, Clone:Hyb-K41220A, Abnova™

Catalog No. 89333387 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-387 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-387 Supplier Abnova Corporation Supplier No. MAB16763
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant human HSP90AA1/HSP90AB1.

Specifications

Antigen HSP90AA1/HSP90AB1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone Hyb-K41220A
Conjugate Biotin
Description heat shock protein 90kDa alpha (cytosolic), class A member 1
Dilution ELISA
Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.09% sodium azide)
Gene HSP90AA1
Gene Alias FLJ31884, HSP86, HSP89A, HSP90A, HSP90N, HSPC1, HSPCA, HSPCAL1, HSPCAL4, HSPN, Hsp89, Hsp90, LAP2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 291-304 of human HSP90 alpha.
Quantity 100 μg
Gene ID (Entrez) 3320, 3326
Target Species Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.