Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90B1 Rabbit anti-Human, Unconjugated, Clone: HSP90B1/3168R, Abnova™

Catalog No. 89339663 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-663 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-663 Supplier Abnova Corporation Supplier No. RAB00648
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

HSP90B1 Monoclonal antibody specifically detects HSP90B1 in Bovine, Chicken, Clawed Frog, Dog, Guinea pig, Hamster, Horse, Human, Monkey, Mouse, Pig, Rat, Sheep samples. It is validated for IHC-P, Flow Cyt.

Specifications

Antigen HSP90B1
Applications Immunohistochemistry (Paraffin), Flow Cytometry
Classification Monoclonal
Clone HSP90B1/3168R
Conjugate Unconjugated
Description heat shock protein 90kDa beta (Grp94), member 1
Gene HSP90B1
Gene Alias ECGP, GP96, GRP94, TRA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to full-length human HSP90B1
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7184
Target Species Bovine, Chicken, Frog, Dog, Guinea Pig, Hamster, Equine, Human, Monkey, Mouse, Pig, Rat, Sheep
Content And Storage Store at -20 to -80°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.