Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP90B1 Rat anti-Chicken, Human, Clone: 9G10.F8.2, Abnova™

Catalog No. 89934302 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-302 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-302 Supplier Abnova Corporation Supplier No. MAB13445
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native chicken HSP90B1.

Specifications

Antigen HSP90B1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 9G10.F8.2
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native chicken HSP90B1.
Dilution Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be dete
Formulation In 10mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Alias hsp108
Gene Symbols HSP90B1
Host Species Rat
Immunogen Native purified HSP90B1 from chicken oviducts.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374163
Test Specificity This monoclonal antibody is highly specific to HSP90B1 and shows minimal cross-reaction with other members of the HSP90 family.
Target Species Chicken, Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.