Learn More
Invitrogen™ HSP90B2P Polyclonal Antibody

Description
The antibody detects endogenous level of total GRP94B / TRAP1 protein.
Specifications
Specifications
| Antigen | HSP90B2P |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | HSP90B2P |
| Gene Accession No. | Q58FF3 |
| Gene Alias | GRP94b; GRP94P1; Heat shock protein; heat shock protein 90 beta family member 2, pseudogene; HSP; HSP90B2P; HSPCP2; TRA1P1; TRAP1 |
| Gene Symbols | HSP90B2P |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.