Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPA12A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA526768

Catalog No. PIPA526768


Only null left
Add to Cart

Description

Description

HSPA12A Polyclonal Antibody for Western Blot, IHC (P)

HSPA12A is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSPA12A
Polyclonal
Unconjugated
HSPA12A
1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A; Heat shock protein; heat shock protein 12A; heat shock protein family A (Hsp70) member 12A; HSP; HSPA12A; KIAA0417; mKIAA0417
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
259217, 73442
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
O43301, Q8K0U4
HSPA12A
KLH conjugated synthetic peptide between 398-427 amino acids from the central region of human HSPA12A.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.