Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPA12A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567279
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567279 Supplier Invitrogen™ Supplier No. PA567279
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADKEAGGSDG PRETAPTSAY SSPARSLGDT GITPLSPSHI VNDTDSNVSE QQSFLVVVAV Highest antigen sequence identity to the following orthologs - mouse 85%, rat 87%.

HSPA12A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA12A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSPA12A
Gene Accession No. O43301
Gene Alias 1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A; Heat shock protein; heat shock protein 12A; heat shock protein family A (Hsp70) member 12A; HSP; HSPA12A; KIAA0417; mKIAA0417
Gene Symbols HSPA12A
Host Species Rabbit
Immunogen Recombinant Human HSPA12A. Recombinant protein control fragment (Product #RP-107916).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259217
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.