Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPA12A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597845 Supplier Invitrogen™ Supplier No. PA597845
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HSPA12A Polyclonal Antibody for Western Blot, ELISA

HSPA12A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA12A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HSPA12A
Gene Accession No. O43301
Gene Alias 1700063D12Rik; AI118035; AI840429; AW048913; AW556406; D5Mgi40; heat shock 70 kDa protein 12A; heat shock 70kD protein 12A; heat shock 70kDa protein 12A; Heat shock protein; heat shock protein 12A; heat shock protein family A (Hsp70) member 12A; HSP; HSPA12A; KIAA0417; mKIAA0417
Gene Symbols HSPA12A
Host Species Rabbit
Immunogen Recombinant Human Heat shock 70 kDa protein 12A protein (2-231AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259217
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.