Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPA12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553076
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553076 Supplier Invitrogen™ Supplier No. PA553076
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QLLDLSGRAP GGGRLGERRS IDSSFRQARE QLRRSRHSRT FLVESGVGEL WAEMQAGDRY VVA Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

The protein encoded by this gene contains an atypical heat shock protein 70 (Hsp70) ATPase domain and is therefore a distant member of the mammalian Hsp70 family. This gene may be involved in susceptibility to atherosclerosis. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Dec 2015].
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA12B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSPA12B
Gene Accession No. Q96MM6, Q9CZJ2
Gene Alias 2700081N06Rik; C20orf60; Heat shock 70 kDa protein 12B; Heat shock protein; heat shock protein 12B; heat shock protein 70kDa 12B; heat shock protein family A (Hsp70) member 12B; HSP; HSPA12B
Gene Symbols HSPA12B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HSPA12B. Recombinant protein control fragment (Product #RP-90758).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116835, 311427, 72630
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.