Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspA12B Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305107100UL
Description
HspA12B Polyclonal specifically detects HspA12B in Human samples. It is validated for Western Blot.Specifications
HspA12B | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
116835 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
C20orf60, chromosome 20 open reading frame 60, dJ1009E24.2, FLJ32150, heat shock 70 kDa protein 12B, heat shock 70kD protein 12B, MGC131912 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 552-686 of human HSPA12B (NP_443202.3). | |
100 μL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction