Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA14 Rabbit anti-Human, Clone: SD08-48, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP267827
Description
HSPA14 Monoclonal specifically detects HSPA14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HSPA14 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:50-1:200 | |
heat shock 70kDa protein 14, HSP70-4, HSP70L1 | |
Rabbit | |
Protein A purified | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
SD08-48 | |
Unconjugated | |
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide | |
HSPA14 | |
Synthetic peptide within Human HSPA14 aa 51-100/ 509. (SwissProt: Q0VDF9 Human; SwissProt: Q99M31 Mouse; SwissProt: Q6AYB4 Rat) | |
100 μL | |
51182 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction