Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA14 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572469
Description
The heat shock proteins (HSPs) comprise a group of highly conserved, abundantly expressed proteins with diverse functions, including the assembly and sequestering of multi-protein complexes, the transportation of nascent poly-peptide chains across cellular membranes and the regulation of protein folding. HSPA14 (heat shock 70kDa protein 14), also known as HSP70-4 or HSP70L1, is a 509 amino acid novel HSP protein derived from human dendritic cells. Belonging to the heat shock protein 70 family, HSPA14 is thought to promote dendritic cell maturation. It is also suggested that HSPA14 stimulates secretion of the proinflammatory cytokines interleukin 12p70 (IL-12p70), IL-1 beta, TNF alpha, and the chemokines IP-10, MIP-1 alpha, MIP-1 beta and normal T cell expressed and secreted (RANTES).
Specifications
HSPA14 | |
Polyclonal | |
Unconjugated | |
HSPA14 | |
Heat shock 70 kDa protein 14; HSP70-like protein 1Heat shock protein HSP60; HSPA14; HSP60; HSP70L1; HSP7E | |
Rabbit | |
Protein A | |
RUO | |
50497, 51182 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.77 mg/mL | |
PBS with 0.09% sodium azide | |
Q0VDF9, Q99M31 | |
HSPA14 | |
KLH conjugated synthetic peptide between 80-109 amino acids from the N-terminal region of human Hsp 60 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction