Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPA1A Monoclonal Antibody (C9)
GREENER_CHOICE

Catalog No. PIMA541597
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541597 200 μL
PIMA543777 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541597 Supplier Invitrogen™ Supplier No. MA541597
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

HSPA1A Monoclonal Antibody for Western Blot

This intronless gene encodes a 70kDa heat shock protein which is a member of the heat shock protein 70 family. In conjuction with other heat shock proteins, this protein stabilizes existing proteins against aggregation and mediates the folding of newly translated proteins in the cytosol and in organelles. It is also involved in the ubiquitin-proteasome pathway through interaction with the AU-rich element RNA-binding protein 1. The gene is located in the major histocompatibility complex class III region, in a cluster with two closely related genes which encode similar proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA1A
Applications Western Blot
Classification Monoclonal
Clone C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene HSPA1A
Gene Accession No. P0DMV8, P0DMW0, Q27975, Q61696
Gene Alias 68 kDa heat shock protein; APG2; APG-2; dnaK-type molecular chaperone HSP70-1; epididymis secretory protein Li 103; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 1; heat shock 70 kDa protein 1/2; heat shock 70 kDa protein 1A; heat shock 70 kDa protein 1A/1B; Heat shock 70 kDa protein 1B; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70 kDa protein 4; heat shock 70kD protein 1A; heat shock 70kD protein 4; heat shock 70kDa protein 1A; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; heat shock protein 1A; heat shock protein 70; heat shock protein 70-1; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 1A; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; heat shock protein, 70 kDa 3; heat shock-induced protein; HEL-S-103; HEL-S-5a; HS24/P52; hsp68; hsp70; hsp70 RY; HSP70.1; HSP70.1/2; HSP70.1/HSP70.2; Hsp70.3; HSP70-1; HSP70-1/HSP70-2; HSP70-1A; Hsp70-2; Hsp70-3; Hsp70A1; HSP70I; hsp70RY; HSP72; Hspa1; HSPA1A; Hspa1b; Hspa2; HSPA4; HSPH2; HSX70; inducible heat shock protein 70; RY
Gene Symbols HSPA1A
Host Species Mouse
Immunogen Recombinant protein Heat Shock 70kDa Protein 1A. The antigen corresponds to amino acid range 285-641 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 193740, 24472, 282254, 3303
Target Species Human, Mouse, Rat, Canine, Rabbit, Bovine, Pig, Chicken, Guinea Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.