Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspA1B Antibody (HSPA1B/7624), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320484100UG
Description
HspA1B Monoclonal antibody specifically detects HspA1B in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
HspA1B | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
FLJ54328, Heat shock 70 kDa protein 1/2, heat shock 70 kDa protein 1A/1B, heat shock 70kD protein 1B, heat shock 70kDa protein 1B, HSP70.1/HSP70.2, HSP70-1/HSP70-2, HSP70-1B, HSP70-2, HSPA1, HSPA1A | |
Recombinant fragment (around aa400-600) of human HspA1B protein (exact sequence is proprietary) | |
100 μg | |
Cancer | |
3304 | |
Store at 4&drg;C. Do not freeze. | |
IgG2b κ |
Immunohistochemistry (Paraffin) | |
HSPA1B/7624 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction