Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspA1L Mouse anti-Human, Mouse, Rat, Clone: 7D1, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP275530
Description
HspA1L Monoclonal specifically detects HspA1L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HspA1L | |
Monoclonal | |
2 mg/mL | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin | |
heat shock 10kDa protein 1-like, Heat shock 70 kDa protein 1-Hom, Heat shock 70 kDa protein 1L, heat shock 70 kDa protein 1-like, heat shock 70kD protein-like 1, heat shock 70kDa protein 1-like, HSP70-1L, HSP70-HOM, HSP70T, hum70t | |
Mouse | |
70 kDa | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
7D1 | |
Unconjugated | |
PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide | |
HSPA1L | |
Recombinant protein with Human HspA1L aa 335-639/ 641. (SwissProt: P34931 Human; SwissProt: P16627 Mouse; SwissProt: P55063 Rat) | |
Immunogen affinity purified | |
Cancer | |
3305.0 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction