Learn More
Invitrogen™ HSPA2 Polyclonal Antibody

Description
Recommended positive controls: A549, H1299, HCT116, MCF-7, NIH-3T3, BCL-1, U87-MG, SK-N-SH, IMR32, SK-N-AS, PC-12, Rat-2, rat brain. Predicted reactivity: Mouse (99%), Rat (98%), Xenopus laevis (92%), Chicken (96%), Chimpanzee (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | HSPA2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | HSPA2 |
| Gene Accession No. | P14659, P17156, P54652 |
| Gene Alias | 70 kDa heat shock protein; 70kDa; Hcp70.2; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 2; heat shock 70kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein 70.2; heat shock protein alpha 2; heat shock protein family A (Hsp70) member 2; heat shock protein, 70 kDa 2; heat shock-related 70 kDa protein 2; HSP; HSP70.2; HSP70.3; Hsp70-2; HSP70-3; HSP70A2; Hspa2; Hspt70; HST; Hst70; LOW QUALITY PROTEIN: heat shock-related 70 kDa protein 2; Testis-specific heat shock protein-related; testis-specific heat shock protein-related gene hst70 |
| Gene Symbols | HSPA2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.