Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581895 Supplier Invitrogen™ Supplier No. PA581895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILNVTAADKS TGKENKITIT NDKGRLSKDD IDRMVQEAER YKSEDEANRD RVAAKNALES YTYNIKQTVE DEKLRGKISE QDKNKILDKC QEVINWLDRN QMAEKDEYEH KQKELERVCN PIISKLYQGG PGGGSGGGGS GASGGPTIEE Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.

In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HSPA2
Gene Accession No. P14659, P17156, P54652
Gene Alias 70 kDa heat shock protein; 70kDa; Hcp70.2; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 2; heat shock 70kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein 70.2; heat shock protein alpha 2; heat shock protein family A (Hsp70) member 2; heat shock protein, 70 kDa 2; heat shock-related 70 kDa protein 2; HSP; HSP70.2; HSP70.3; Hsp70-2; HSP70-3; HSP70A2; Hspa2; Hspt70; HST; Hst70; LOW QUALITY PROTEIN: heat shock-related 70 kDa protein 2; Testis-specific heat shock protein-related; testis-specific heat shock protein-related gene hst70
Gene Symbols HSPA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HSPA2. Recombinant protein control fragment (Product #RP-88859).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15512, 3306, 60460
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.