Learn More
Invitrogen™ HSPA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596853
Description
Immunogen sequence: TPLSLGIETA GGVMTPLIKR NTTIPTKQTQ TFTTYSDNQS SVLVQVYEGE RAMTKDNNLL GKFDLTGIPP APRGVPQIEV TFDIDANGIL NVTAADKSTG KENKITITND KGRLSKDDID RMVQEAERYK SEDEANRDRV AAKNALESYT YNIKQTVEDE KLRGKISEQD KNKILDKCQE VINWLDRNQM AEKDEYEHKQ KELERVCNPI ISKLYQGGPG GGSGGGGSGA SGGPTIEEVD; Positive Samples: MCF7, A-549, SKOV3, HeLa, A-431, Mouse brain, Mouse testis, Rat brain, Rat kidney.
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
Specifications
HSPA2 | |
Polyclonal | |
Unconjugated | |
HSPA2 | |
70 kDa heat shock protein; 70kDa; Hcp70.2; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 2; heat shock 70kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein 70.2; heat shock protein alpha 2; heat shock protein family A (Hsp70) member 2; heat shock protein, 70 kDa 2; heat shock-related 70 kDa protein 2; HSP; HSP70.2; HSP70.3; Hsp70-2; HSP70-3; HSP70A2; Hspa2; Hspt70; HST; Hst70; LOW QUALITY PROTEIN: heat shock-related 70 kDa protein 2; Testis-specific heat shock protein-related; testis-specific heat shock protein-related gene hst70 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
15512, 3306, 60460 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P14659, P17156, P54652 | |
HSPA2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 400-639 of human HSPA2 (NP_0688142). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.