Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117993
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117993 Supplier Invitrogen™ Supplier No. PA5117993
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HSPA2 Polyclonal Antibody for Western Blot, IHC (P), IP

In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene HSPA2
Gene Accession No. P54652
Gene Alias 70 kDa heat shock protein; 70kDa; Hcp70.2; Heat shock 70 kDa protein 2; Heat shock 70 kDa protein 3; heat shock 70kD protein 2; heat shock 70kDa protein 2; Heat shock protein; heat shock protein 2; heat shock protein 70.2; heat shock protein alpha 2; heat shock protein family A (Hsp70) member 2; heat shock protein, 70 kDa 2; heat shock-related 70 kDa protein 2; HSP; HSP70.2; HSP70.3; Hsp70-2; HSP70-3; HSP70A2; Hspa2; Hspt70; HST; Hst70; LOW QUALITY PROTEIN: heat shock-related 70 kDa protein 2; Testis-specific heat shock protein-related; testis-specific heat shock protein-related gene hst70
Gene Symbols HSPA2
Host Species Rabbit
Immunogen E. coli-derived Human HSPA2 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3306
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.