Learn More
HSPA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | HSPA2 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8257
![]() ![]() |
Proteintech Group Inc
127971AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody well recognized the endogenous HSPA4 protein in testis and multiple cell lines.
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.Specifications
HSPA2 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P14659, P17156, P54652 | |
15512, 3306, 60460 | |
HSPA2 Fusion Protein Ag3539 | |
Primary | |
-20°C | |
HSPA2 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Heat shock 70 kDa protein 2, heat shock 70kDa protein 2, HSP70 2, HSP70 3, HSPA2 | |
HSPA2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.