Learn More
Invitrogen™ HSPA4 Monoclonal Antibody (5A6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515578
Description
MA5-15578 targets HSP70 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15578 immunogen is purified recombinant fragment of human HSP70 expressed in E. Coli. MA5-15578 detects HSP70 which has a predicted molecular weight of approximately 70kDa.
The HSP 70 family is composed of four highly conserved proteins: HSP 70, HSC 70, GRP 75 and GRP 78. HSP 70 expression is strongly induced in response to heat stress. HSP 70 and HSC 70 play key roles in the cytosolic endoplasmic reticulum and mitochondrial import machinery and are found in both the cytosol and nucleus of mammalian cells. GRP 78 is localized in the endoplasmic reticulum, where it receives imported secretory proteins and is involved in the folding and translocation of nascent peptide chains. GRP 75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. It has been postulated that members of the HSP 70 family act as force-generating motors, relying on the hydrolysis of ATP for their activity.
Specifications
HSPA4 | |
Monoclonal | |
1.0 mg/mL | |
PBS with 0.05% sodium azide | |
P34932, Q61316 | |
HSPA4 | |
Purified recombinant fragment of human HSP70 expressed in E. Coli. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
5A6 | |
Unconjugated | |
HSPA4 | |
70kDa; AI317151; APG2; APG-2; epididymis secretory sperm binding protein Li 5a; heat shock 70 kDa protein 4; heat shock 70kD protein 4; heat shock 70kDa protein 4; Heat shock 70-related protein APG-2; Heat shock protein; heat shock protein 4; heat shock protein Apg-2; heat shock protein family A (Hsp70) member 4; heat shock protein, 110 kDa; HEL-S-5a; HS24/P52; Hsp110; hsp70; hsp70 RY; Hsp70RY; Hspa4; HSPH2; Irp94; ischemia responsive 94 kDa protein; mKIAA4025; RY | |
Mouse | |
Purified | |
RUO | |
15525, 3308 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.