Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspA4L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | HspA4L |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
HspA4L Polyclonal specifically detects HspA4L in Human samples. It is validated for Western Blot.Specifications
| HspA4L | |
| Polyclonal | |
| Rabbit | |
| O95757 | |
| 22824 | |
| Synthetic peptides corresponding to HSPA4L(heat shock 70kDa protein 4-like) The peptide sequence was selected from the middle region of HSPA4L. Peptide sequence KCHAEHTPEEEIDHTGAKTKSAVSDKQDRLNQTLKKGKVKSIDLPIQSSL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| APG1, APG-1, heat shock 70 kDa protein 4L, heat shock 70 kDa protein 4-like protein, heat shock 70kDa protein 4-like, Heat shock 70-related protein APG-1, heat shock protein (hsp110 family), Osmotic stress protein 94, Osp94 | |
| HSPA4L | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title