Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HSPA4L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142360
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142360 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142360 Supplier Invitrogen™ Supplier No. PA5142360
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

HSPA4L possesses chaperone activity in vitro where it inhibits aggregation of citrate synthase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA4L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene HSPA4L
Gene Accession No. O95757
Gene Alias 94kDa; AI461691; APG1; APG-1; heat shock 70 kDa protein 4L; heat shock 70 kDa protein 4-like protein; heat shock 70 kDa protein 4L-like; Heat shock 70 kDa protein 4L-like protein; heat shock 70kDa protein 4 like; heat shock 70kDa protein 4-like; heat shock 70-related protein APG-1; Heat shock protein; heat shock protein (hsp110 family); heat shock protein 4 like; heat shock protein 4-like; heat shock protein family A (Hsp70) member 4 like; HSP; Hsp4l; HSPA4L; HSPH3; LOW QUALITY PROTEIN: heat shock 70 kDa protein 4L; osmotic stress protein 94; OSP94; testis tissue sperm-binding protein Li 64n
Gene Symbols HSPA4L
Host Species Goat
Immunogen Peptide with sequence C-DSSQHTKSSGEMEVD.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22824
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.