Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA5 Mouse anti-Human, Biotin, Clone:1H11-1H7, Abnova™

Catalog No. 89334123 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-123 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-123 Supplier Abnova Corporation Supplier No. MAB17320
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against human HSPA5.

When Chinese hamster K12 cells are starved of glucose, the synthesis of several proteins, called glucose-regulated proteins (GRPs), is markedly increased. Hendershot et al. (1994) [PubMed 8020977] pointed out that one of these, GRP78 (HSPA5), also referred to as ′immunoglobulin heavy chain-binding protein′ (BiP), is a member of the heat-shock protein-70 (HSP70) family and is involved in the folding and assembly of proteins in the endoplasmic reticulum (ER). Because so many ER proteins interact transiently with GRP78, it may play a key role in monitoring protein transport through the cell.[supplied by OMIM]

Specifications

Antigen HSPA5
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1H11-1H7
Conjugate Biotin
Description heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa)
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Western Blot (1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene HSPA5
Gene Alias BIP, FLJ26106, GRP78, MIF2
Host Species Mouse
Immunogen His-tagged human HSPA5.
Quantity 100 μg
Gene ID (Entrez) 3309
Target Species Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.