Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HspA6 Antibody (OTI1H3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270959MFV500
Description
HspA6 Monoclonal antibody specifically detects HspA6 in Human samples. It is validated for Flow CytometrySpecifications
HspA6 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
3310 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
OTI1H3 | |
Flow Cytometry | |
heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B' | |
Full length human recombinant protein of human HSPA6 (NP_002146) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction