Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA6/HSPA7 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5100823
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100823 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100823 Supplier Thermo Scientific Supplier No. PA5100823

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HSP77/76.

HSPA6/HSPA7 gene ontology (GO) annotations include cellular response to heat; cellular response to unfolded protein; chaperone cofactor-dependent protein refolding; protein refolding; response to unfolded protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HSPA6/HSPA7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HSPA7
Gene Accession No. P17066, P48741
Gene Alias Heat shock 70 kDa protein 6; heat shock 70 kDa protein B'; heat shock 70kD protein 6 (HSP70B'); heat shock 70kDa protein 6 (HSP70B'); heat shock protein family A (Hsp70) member 6; heat shock protein family A (Hsp70) member 7; HSP70B; HSP70B'; HSPA6; HSPA7
Gene Symbols HSPA6, HSPA7
Host Species Rabbit
Immunogen A synthesized peptide derived from human HSPA7(Accession P48741), corresponding to amino acid residues D227-S277.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3310, 3311
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.