Learn More
HSPA6 Mouse anti-Human, Clone: OTI1H3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501773

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.Specifications
HSPA6 | |
Monoclonal | |
Unconjugated | |
HSPA6 | |
HSP70B′ | |
Mouse | |
Affinity Chromatography | |
RUO | |
3310 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P17066 | |
HSPA6 | |
Full length human recombit protein of human HSPA6 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.