Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HSPA6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544195
Description
Peptide sequence: EYEHQKRELE QICRPIFSRL YGGPGVPGGS SCGTQARQGD PSTGPIIEEV Sequence homology: Human: 100%; Pig: 79%.
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.
Specifications
| HSPA6 | |
| Polyclonal | |
| Unconjugated | |
| HSPA6 | |
| heat shock 70 kDa protein; heat shock 70 kDa protein 6; Heat shock 70 kDa protein B'; heat shock 70kD protein 6 (HSP70B'); heat shock 70kDa protein 6 (HSP70B'); Heat shock protein; heat shock protein 70; heat shock protein 70, hsp70; heat shock protein family A (Hsp70) member 6; HSP; HSP70; HSP70B'; HSPA6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3310 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P17066 | |
| HSPA6 | |
| Synthetic peptide directed towards the middle region of human HSPA6. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction