Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA6 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987322
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7322 Supplier Bioss Supplier No. BS6540R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.

Specifications

Antigen HSPA6
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HSPA6
Gene Accession No. P17066
Gene Alias heat shock 70 kDa protein; Heat shock 70 kDa protein 6; Heat shock 70 kDa protein B′; heat shock 70kD protein 6 (HSP70B′); heat shock 70kDa protein 6 (HSP70B′); Heat shock protein; heat shock protein 70; heat shock protein family A (Hsp70) member 6; HSP; HSP70; HSP70B′; HSPA6
Gene Symbols HSPA6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HSPA6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3310
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.