Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA8/HSC71/Hsc70 Antibody (1B5), Novus Biologicals™

Rat Monoclonal Antibody
$214.28 - $776.74
Specifications
| Antigen | HSPA8/HSC71/Hsc70 |
|---|---|
| Clone | 1B5 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Electron Microscopy 1:10-1:500, Enzyme Immunoassay |
| Applications | Immunocytochemistry, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018817
![]() |
Novus Biologicals
NBP1978680.025MG |
0.025 mg |
Each for $214.28
|
|
|||||
NBP19786805
![]() |
Novus Biologicals
NBP1978680.05MG |
0.05 mg |
Each for $443.24
|
|
|||||
NBP19786802
![]() |
Novus Biologicals
NBP1978680.2MG |
0.2 mg |
Each for $776.74
|
|
|||||
Description
HSPA8/HSC71/Hsc70 Monoclonal specifically detects HSPA8/HSC71/Hsc70 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Electron Microscopy.Specifications
| HSPA8/HSC71/Hsc70 | |
| 1.0 mg/mL | |
| Immunocytochemistry, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Pig, Bovine, Hamster, Sheep, Canine, Chicken, Rat, Rabbit, Human, Guinea Pig, Monkey, Mouse | |
| Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 | |
| HSPA8 | |
| IgG2a | |
| Protein G purified | |
| Recognizes dog, chicken, guinea pig, pig, rabbit, sheep, and turtle HSC70 (HSP73). |
| 1B5 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Electron Microscopy 1:10-1:500, Enzyme Immunoassay | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
| 3312 | |
| Native hamster Hsc70 (Hsp73). | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title