Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA8/HSC71/Hsc70 Antibody (OTI1H3), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270962IR
Description
HSPA8/HSC71/Hsc70 Monoclonal specifically detects HSPA8/HSC71/Hsc70 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
HSPA8/HSC71/Hsc70 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HSPA8 | |
Full length recombinant protein of human HSPA8/HSC71/Hsc70 (NP_006588) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG3 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1H3 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 | |
Mouse | |
Affinity Purified | |
Membrane Trafficking and Chaperones | |
3312.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction