Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSPA8/HSC71/Hsc70 Rabbit anti-Human, Mouse, Rat, Bovine, Hamster, Monkey, Primate, Rabbit, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB018072 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018072 0.025 mg
NB120142701 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018072 Supplier Novus Biologicals Supplier No. NB12014270.025MG

Rabbit Polyclonal Antibody

HSPA8/HSC71/Hsc70 Polyclonal specifically detects HSPA8/HSC71/Hsc70 in Human, Mouse, Rat, Bovine, Hamster, Monkey, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen HSPA8/HSC71/Hsc70
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry
Formulation PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Gene Accession No. P11142
Gene Alias Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71
Gene Symbols HSPA8
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 559 of human Hsc70.
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 3312
Test Specificity The detects a 71 kDa Hsc70. A ∼40 kDa band can also be detected in human samples.
Target Species Primate, Human, Hamster, Monkey, Bovine, Rabbit, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.