Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSPA8/HSC71/Hsc70 Rabbit anti-Human, Mouse, Rat, Clone: 0J2Z9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | HSPA8/HSC71/Hsc70 |
---|---|
Clone | 0J2Z9 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
HSPA8/HSC71/Hsc70 Monoclonal antibody specifically detects HSPA8/HSC71/Hsc70 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
HSPA8/HSC71/Hsc70 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Membrane Trafficking and Chaperones | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3312 | |
IgG | |
Affinity purified |
0J2Z9 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Heat shock 70 kDa protein 8, heat shock 70kD protein 8, heat shock 70kDa protein 8, heat shock cognate 71 kDa protein, heat shock cognate protein 54, heat shock cognate protein, 71-kDa, HSC54, HSC70heat shock 70kd protein 10, HSC71, HSP71, HSP73constitutive heat shock protein 70, HSPA10, LAP1, lipopolysaccharide-associated protein 1, LPS-associated protein 1, MGC131511, MGC29929, NIP71, N-myristoyltransferase inhibitor protein 71 | |
A synthetic peptide corresponding to a sequence within amino acids 547-646 of human HSPA8/HSC71/Hsc70 (P11142). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title