Learn More
HSPA9/GRP75/Mortalin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | HSPA9/GRP75/Mortalin |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-9944
![]() |
Bethyl Laboratories, Inc
A305256AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Orangutan
The HSP70 family is composed of four highly conserved proteins: HSP70, HSC70, GRP75 and GRP78. These proteins serve a variety of roles. GRP75 expression is restricted to the mitochondrial matrix and aids in the translocation and folding of nascent polypeptide chains of both nuclear and mitochondrial origin. GRP75 and GRP78 are unresponsive to heat stress and are induced by glucose deprivation.Specifications
HSPA9/GRP75/Mortalin | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CSA; GRP-75; GRP75; HSPA9B; MOT; MOT2; MTHSP75; PBP74; 75 kDa glucose-regulated protein; heat shock 70kD protein 9B; mortalin; perinuclear; mortalin-2; p66-mortalin; peptide-binding protein 74; stress-70 protein; mitochondrial | |
Between 629 and 679 | |
Primary | |
4° C | |
HSPA9 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P38646, P38647 | |
15526, 3313 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.